siRNA Details
| Gene Symbol: BIVM-ERCC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204425.1 | NP_001191354.1 | 15 | 2198 | ||
| GenBank mRNA | AB209312.1 | - | 15 | 1990 | ||
| LM994822.1 | - | 15 | 2076 | |||
| Gene Symbol: ERCC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000123.3 | NP_000114.2 | 7 | 1140 | ||
| GenBank mRNA | AF462447.1 | - | 7 | 1097 | ||
| AK294708.1 | - | 7 | 1069 | |||
| AK299758.1 | - | 7 | 852 | |||
| AK303558.1 | - | 8 | 1043 | |||
| BC031522.2 | - | 7 | 1126 | 3869893 | ||
| BX647399.1 | - | 7 | 969 | |||
| D16305.1 | - | 7 | 724 | |||
| L20046.1 | - | 7 | 762 | |||
| LM994821.1 | - | 3 | 210 | |||
| X69978.1 | - | 7 | 911 | |||

BIVM-ERCC5
Genomic Map