siRNA Details
| Gene Symbol: F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000128.3 | NP_000119.1 | 14 | 1992 | ||
| XM_005262821.3 | XP_005262878.1 | 14 | 2014 | |||
| XM_005262822.3 | XP_005262879.1 | 13 | 1918 | |||
| XM_005262823.3 | XP_005262880.1 | 12 | 1744 | |||
| XM_006714137.2 | XP_006714200.1 | 14 | 1966 | |||
| GenBank mRNA | AF045649.1 | - | 13 | 1531 | ||
| AK313195.1 | - | 14 | 1766 | |||
| BC119014.1 | - | 14 | 1755 | 40085167 | ||
| BC122863.1 | - | 14 | 1755 | 40085169 | ||
| M13142.1 | - | 14 | 1702 | |||

F11
Genomic Map