siRNA Details
| Gene Symbol: 2410127L17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026120.4 | NP_080396.2 | 5 | 1081 | ||
| GenBank mRNA | AK010784.1 | - | 5 | 966 | ||
| AK082071.1 | - | 1 | 2985 | |||
| BC019559.1 | - | 5 | 1271 | |||
| BC043335.1 | - | 5 | 1035 | 5030742 | ||
| BC092281.1 | - | 5 | 913 | 4950426 | ||

2410127L17Rik
Genomic Map