siRNA Details
| Gene Symbol: Txndc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026132.2 | NP_080408.2 | 3, 4 | 292 | ||
| GenBank mRNA | AK006683.1 | - | 3, 4 | 272 | ||
| AK015240.1 | - | 3, 4 | 293 | |||
| AY495589.1 | - | 3, 4 | 197 | |||
| BC049564.1 | - | 3, 4 | 263 | |||
| BC116763.1 | - | 3, 4 | 293 | 40126082 | ||
| BC116765.1 | - | 3, 4 | 293 | 40126084 | ||

Txndc8
Genomic Map