siRNA Details
| Gene Symbol: 4930449I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026136.2 | NP_080412.1 | 921 | |||
| GenBank mRNA | AK015434.1 | - | 922 | |||
| BC147588.1 | - | 918 | 9056578 | |||
| BC147604.1 | - | 918 | 9056594 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930449I24Rik| Gene Symbol: 4930449I24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026136.2 | NP_080412.1 | 921 | |||
| GenBank mRNA | AK015434.1 | - | 922 | |||
| BC147588.1 | - | 918 | 9056578 | |||
| BC147604.1 | - | 918 | 9056594 | |||