siRNA Details
| Gene Symbol: Mgat4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162368.1 | NP_001155840.1 | 5 | 478 | ||
| NM_001162369.1 | NP_001155841.1 | 5 | 351 | |||
| NM_001205098.1 | NP_001192027.1 | 7 | 891 | |||
| NM_026243.4 | NP_080519.2 | 3 | 373 | |||
| GenBank mRNA | AK018574.1 | - | 3 | 372 | ||
| AK018672.1 | - | 3 | 376 | |||
| AK032009.1 | - | 5 | 352 | |||
| AK033482.1 | - | 3 | 374 | |||
| BB611341.1 | - | 5 | 479 | |||
| BC046987.1 | - | 3 | 361 | 6475851 | ||
| CD774691.1 | - | 7 | 659 | |||

Mgat4c
Genomic Map