siRNA Details
| Gene Symbol: Gin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026250.2 | NP_080526.2 | 3 | 296 | ||
| GenBank mRNA | AK014287.1 | - | 3 | 308 | ||
| AK015243.1 | - | 3 | 275 | |||
| AK018404.1 | - | 3 | 297 | |||
| AK084620.1 | - | 2 | 200 | |||
| AK090002.1 | - | 3 | 281 | |||
| AK169361.1 | - | 3 | 259 | |||
| BC033572.1 | - | 2 | 241 | 5067692 | ||

Gin1
Genomic Map