siRNA Details
| Gene Symbol: 4930524B15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026262.1 | NP_080538.1 | 1, 2 | 327 | ||
| GenBank mRNA | AK015006.1 | - | 1, 2 | 369 | ||
| AK015879.1 | - | 1, 2 | 327 | |||
| BC114574.1 | - | 1, 2 | 317 | 40049349 | ||

4930524B15Rik
Genomic Map