siRNA Details
| Gene Symbol: Narf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026272.3 | NP_080548.3 | 2, 3 | 211 | ||
| GenBank mRNA | AK013432.1 | - | 2, 3 | 170 | ||
| AK076232.1 | - | 2, 3 | 194 | |||
| AK081305.1 | - | 2, 3 | 195 | |||
| BC016090.1 | - | 2, 3 | 150 | 4503713 | ||
| BM951066.1 | - | 2, 3 | 211 | |||

Narf
Genomic Map