siRNA Details
| Gene Symbol: Samd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026283.2 | NP_080559.1 | 1, 2 | 258 | ||
| GenBank mRNA | AK006211.1 | - | 1, 2 | 259 | ||
| AK015593.1 | - | 1, 2 | 237 | |||
| AK075749.1 | - | 1, 2 | 209 | |||
| BC116745.1 | - | 1, 2 | 246 | 40126064 | ||
| BC116747.1 | - | 1, 2 | 246 | 40126066 | ||

Samd8
Genomic Map