siRNA Details
| Gene Symbol: Spaca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290443.1 | NP_001277372.1 | 1, 2 | 308 | ||
| NM_026293.3 | NP_080569.1 | 1, 2 | 310 | |||
| GenBank mRNA | AK006187.1 | - | 1, 2 | 311 | ||
| AK007270.1 | - | 1, 2 | 282 | |||
| AK016011.1 | - | 1, 2 | 303 | |||
| BC147079.1 | - | 1, 2 | 307 | 9056040 | ||
| BC147080.1 | - | 1, 2 | 307 | 9056039 | ||
| BY706285.1 | - | 1, 2 | 309 | |||

Spaca1
Genomic Map