siRNA Details
| Gene Symbol: 4930549C01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026300.2 | NP_080576.1 | 1, 2 | 259 | ||
| GenBank mRNA | AK015053.1 | - | 1, 2 | 222 | ||
| AK016074.1 | - | 1, 2 | 260 | |||
| BC048442.1 | - | 1, 2 | 249 | 6704947 | ||

4930549C01Rik
Genomic Map