siRNA Details
| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281871.1 | NP_001268800.1 | 4 | 314 | ||
| NM_026312.5 | NP_080588.1 | 4 | 314 | |||
| GenBank mRNA | AK007655.1 | - | 4 | 296 | ||
| AK007872.1 | - | 3 | 183 | |||
| AK011610.1 | - | 4 | 278 | |||
| AK030507.1 | - | 4 | 296 | |||
| AK076889.1 | - | 4 | 296 | |||
| AK157432.1 | - | 2 | 2154 | |||
| BC035042.1 | - | 4 | 278 | 1224959 | ||
| BC044719.1 | - | 4 | 262 | 6394829 | ||
| BC061070.1 | - | 4 | 275 | 6594922 | ||

Pbdc1
Genomic Map