siRNA Details
| Gene Symbol: Aldh3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026316.2 | NP_080592.2 | 8, 9 | 1146 | ||
| GenBank mRNA | AF362571.1 | - | 7, 8 | 1068 | ||
| AK004004.1 | - | 1, 2 | 165 | |||
| AK005615.1 | - | 2, 3 | 276 | |||
| AK154500.1 | - | 8, 9 | 1192 | |||
| AK154723.1 | - | 8, 9 | 1239 | |||
| AK159447.1 | - | 8, 9 | 1160 | |||
| BC023305.1 | - | 8, 9 | 1167 | |||
| BC046597.1 | - | 8, 9 | 1146 | 6312211 | ||
| BC082792.1 | - | 1, 2 | 184 | |||

Aldh3b1
Genomic Map