siRNA Details
| Gene Symbol: FUT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000149.3 | NP_000140.1 | 3 | 2364 | ||
| NM_001097639.1 | NP_001091108.1 | 3 | 2016 | |||
| NM_001097640.1 | NP_001091109.1 | 3 | 1962 | |||
| NM_001097641.1 | NP_001091110.1 | 2 | 1823 | |||
| XM_011527865.1 | XP_011526167.1 | 3 | 2342 | |||
| XM_011527866.1 | XP_011526168.1 | 3 | 2003 | |||
| XM_011527867.1 | XP_011526169.1 | 3 | 1972 | |||
| GenBank mRNA | AF131913.1 | - | 2 | 1895 | ||
| U27326.1 | - | 3 | 1955 | |||
| U27327.1 | - | 3 | 2009 | |||
| U27328.1 | - | 4 | 2358 | |||
| X53578.1 | - | 2 | 1798 | |||

FUT3
Genomic Map