siRNA Details
| Gene Symbol: Lce1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026335.2 | NP_080611.1 | 2 | 522 | ||
| GenBank mRNA | AK009424.1 | - | 2 | 522 | ||
| AK010064.1 | - | 2 | 523 | |||
| BC117062.1 | - | 483 | ||||
| BC119356.1 | - | 483 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lce1h| Gene Symbol: Lce1h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026335.2 | NP_080611.1 | 2 | 522 | ||
| GenBank mRNA | AK009424.1 | - | 2 | 522 | ||
| AK010064.1 | - | 2 | 523 | |||
| BC117062.1 | - | 483 | ||||
| BC119356.1 | - | 483 | ||||