siRNA Details
| Gene Symbol: Pabpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163836.1 | NP_001157308.1 | 1 | 506 | ||
| GenBank mRNA | AK016578.1 | - | 1 | 499 | ||
| AK029765.1 | - | 1 | 487 | |||
| BC113177.1 | - | 1 | 476 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpc6| Gene Symbol: Pabpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163836.1 | NP_001157308.1 | 1 | 506 | ||
| GenBank mRNA | AK016578.1 | - | 1 | 499 | ||
| AK029765.1 | - | 1 | 487 | |||
| BC113177.1 | - | 1 | 476 | |||