siRNA Details
| Gene Symbol: Gramd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026240.2 | NP_080516.2 | 4, 5 | 528 | ||
| GenBank mRNA | AK018551.1 | - | 5, 6 | 778 | ||
| AK018691.1 | - | 4, 5 | 514 | |||
| AK038289.1 | - | 3, 4 | 569 | |||
| AK161329.1 | - | 4, 5 | 461 | |||
| AK164050.1 | - | 4, 5 | 513 | |||
| BC057978.1 | - | 4, 5 | 503 | 5044170 | ||

Gramd3
Genomic Map