siRNA Details
| Gene Symbol: Mgat4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162368.1 | NP_001155840.1 | 5 | 497 | ||
| NM_001162369.1 | NP_001155841.1 | 5 | 370 | |||
| NM_001205098.1 | NP_001192027.1 | 7 | 910 | |||
| NM_026243.4 | NP_080519.2 | 3 | 392 | |||
| GenBank mRNA | AK018574.1 | - | 3 | 391 | ||
| AK018672.1 | - | 3 | 395 | |||
| AK032009.1 | - | 5 | 371 | |||
| AK033482.1 | - | 3 | 393 | |||
| BB611341.1 | - | 5 | 498 | |||
| BC046987.1 | - | 3 | 380 | 6475851 | ||
| CD774691.1 | - | 7 | 678 | |||

Mgat4c
Genomic Map