siRNA Details
| Gene Symbol: 4930430A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026248.3 | NP_080524.2 | 1, 2 | 430 | ||
| GenBank mRNA | AK015247.1 | - | 1, 2 | 429 | ||
| AK016868.1 | - | 1, 2 | 433 | |||
| BC125324.1 | - | 1, 2 | 381 | 40129839 | ||

4930430A15Rik
Genomic Map