siRNA Details
| Gene Symbol: Ctdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026295.2 | NP_080571.2 | 11 | 2658 | ||
| GenBank mRNA | AK016213.1 | - | 2 | 180 | ||
| AK156617.1 | - | 11 | 2687 | |||
| BC052934.1 | - | 2 | 263 | |||
| BC053435.1 | - | 11 | 2658 | 6394578 | ||

Ctdp1
Genomic Map