siRNA Details
| Gene Symbol: Rabl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042499.1 | NP_001035964.1 | 7, 8 | 666 | ||
| GenBank mRNA | AK011280.1 | - | 7, 8 | 608 | ||
| AK016099.1 | - | 7, 8 | 675 | |||
| AK028195.1 | - | 7, 8 | 660 | |||
| AK028269.1 | - | 7, 8 | 653 | |||
| BC050194.1 | - | 7, 8 | 647 | 30022531 | ||

Rabl3
Genomic Map