siRNA Details
| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281871.1 | NP_001268800.1 | 5 | 394 | ||
| NM_026312.5 | NP_080588.1 | 5 | 394 | |||
| GenBank mRNA | AK007655.1 | - | 5 | 376 | ||
| AK007872.1 | - | 4 | 263 | |||
| AK011610.1 | - | 5 | 358 | |||
| AK030507.1 | - | 5 | 376 | |||
| AK076889.1 | - | 5 | 376 | |||
| AK157432.1 | - | 2 | 2849 | |||
| BC035042.1 | - | 5 | 358 | 1224959 | ||
| BC044719.1 | - | 5 | 342 | 6394829 | ||
| BC061070.1 | - | 5 | 355 | 6594922 | ||

Pbdc1
Genomic Map