siRNA Details
| Gene Symbol: GLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000169.2 | NP_000160.1 | 2 | 356 | ||
| GenBank mRNA | AK222627.1 | - | 2 | 268 | ||
| AK291095.1 | - | 2 | 275 | |||
| AK297148.1 | - | 2 | 268 | |||
| BC002689.2 | - | 2 | 252 | 3609235 | ||
| BT006864.1 | - | 2 | 246 | |||
| D00039.1 | - | 2 | 170 | |||
| X05790.1 | - | 2 | 269 | |||
| X16889.1 | - | 2 | 248 | |||

GLA
Genomic Map