siRNA Details
| Gene Symbol: Aldh3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026316.2 | NP_080592.2 | 10 | 1611 | ||
| GenBank mRNA | AF362571.1 | - | 9 | 1533 | ||
| AK004004.1 | - | 3 | 630 | |||
| AK005615.1 | - | 4 | 741 | |||
| AK154500.1 | - | 10 | 1657 | |||
| AK159447.1 | - | 10 | 1625 | |||
| AK160106.1 | - | 10 | 1626 | |||
| BC023305.1 | - | 10 | 1632 | |||
| BC046597.1 | - | 10 | 1611 | 6312211 | ||
| BC082792.1 | - | 3 | 649 | |||

Aldh3b1
Genomic Map