siRNA Details
| Gene Symbol: Tmem179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026325.3 | NP_080601.2 | 2, 3 | 331 | ||
| GenBank mRNA | AK005135.1 | - | 2, 3 | 250 | ||
| AK007906.1 | - | 2, 3 | 307 | |||
| AK010996.1 | - | 2, 3 | 312 | |||
| BC061068.1 | - | 2, 3 | 231 | |||
| BC087914.1 | - | 2, 3 | 247 | |||
| BC116961.1 | - | 2, 3 | 331 | 40126280 | ||
| BC116963.1 | - | 2, 3 | 331 | 40126282 | ||
| BC144813.1 | - | 2, 3 | 261 | |||

Tmem179b
Genomic Map