siRNA Details
| Gene Symbol: Pcnxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026327.3 | NP_080603.2 | 2 | 477 | ||
| GenBank mRNA | AK007829.1 | - | 2 | 458 | ||
| AK029996.1 | - | 2 | 434 | |||
| AK076035.1 | - | 2 | 478 | |||
| AK136802.1 | - | 2 | 452 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcnxl4| Gene Symbol: Pcnxl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026327.3 | NP_080603.2 | 2 | 477 | ||
| GenBank mRNA | AK007829.1 | - | 2 | 458 | ||
| AK029996.1 | - | 2 | 434 | |||
| AK076035.1 | - | 2 | 478 | |||
| AK136802.1 | - | 2 | 452 | |||