siRNA Details
| Gene Symbol: Fam114a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168667.1 | NP_001162138.1 | 2, 3 | 274 | ||
| NM_026342.3 | NP_080618.1 | 2, 3 | 277 | |||
| GenBank mRNA | AK007968.1 | - | 2, 3 | 251 | ||
| AK018570.1 | - | 2, 3 | 250 | |||
| AK076316.1 | - | 2, 3 | 248 | |||
| AK149763.1 | - | 2, 3 | 252 | |||
| BY134675.1 | - | 2, 3 | 274 | |||

Fam114a2
Genomic Map