siRNA Details
| Gene Symbol: Gpr89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026229.4 | NP_080505.1 | 9, 10 | 899 | ||
| GenBank mRNA | AK009734.1 | - | 8, 9 | 778 | ||
| AK016787.1 | - | 9, 10 | 899 | |||
| BC010729.1 | - | 9, 10 | 892 | 3708143 | ||
More Information
Set Membership: |
|

Gpr89
Genomic Map