siRNA Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026231.2 | NP_080507.2 | 12 | 2464 | ||
| GenBank mRNA | AK013967.1 | - | 9 | 1316 | ||
| AK017059.1 | - | 10 | 1739 | |||
| AK020021.1 | - | 11 | 2074 | |||
| AK033409.1 | - | 12 | 2503 | |||
| BC047136.1 | - | 12 | 2464 | 6311866 | ||

Gstcd
Genomic Map