siRNA Details
| Gene Symbol: Mgat4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026233.2 | NP_080509.2 | 11 | 1200 | ||
| GenBank mRNA | AK016272.1 | - | 11 | 1209 | ||
| AK017057.1 | - | 8 | 790 | |||
| HM067443.1 | - | 11 | 1324 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgat4d| Gene Symbol: Mgat4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026233.2 | NP_080509.2 | 11 | 1200 | ||
| GenBank mRNA | AK016272.1 | - | 11 | 1209 | ||
| AK017057.1 | - | 8 | 790 | |||
| HM067443.1 | - | 11 | 1324 | |||