siRNA Details
| Gene Symbol: 4930524B15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026262.1 | NP_080538.1 | 5 | 807 | ||
| GenBank mRNA | AK015006.1 | - | 5 | 849 | ||
| AK015777.1 | - | 4 | 435 | |||
| AK015879.1 | - | 5 | 807 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930524B15Rik| Gene Symbol: 4930524B15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026262.1 | NP_080538.1 | 5 | 807 | ||
| GenBank mRNA | AK015006.1 | - | 5 | 849 | ||
| AK015777.1 | - | 4 | 435 | |||
| AK015879.1 | - | 5 | 807 | |||