siRNA Details
| Gene Symbol: Ube2r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026275.4 | NP_080551.1 | 4 | 523 | ||
| GenBank mRNA | AJ240086.1 | - | 4 | 482 | ||
| AK003550.1 | - | 4 | 543 | |||
| AK007517.1 | - | 4 | 657 | |||
| AK075703.1 | - | 4 | 533 | |||
| AK075714.1 | - | 4 | 524 | |||
| AK182726.1 | - | 1 | 118 | |||
| AK183933.1 | - | 1 | 118 | |||
| AK217134.1 | - | 1 | 118 | |||
| BC011112.1 | - | 4 | 531 | 3982808 | ||

Ube2r2
Genomic Map