siRNA Details
| Gene Symbol: 4930544G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161773.1 | NP_001155245.1 | 1038 | |||
| GenBank mRNA | AK006363.1 | - | 1 | 998 | ||
| AK016035.1 | - | 1 | 1000 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930544G11Rik| Gene Symbol: 4930544G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161773.1 | NP_001155245.1 | 1038 | |||
| GenBank mRNA | AK006363.1 | - | 1 | 998 | ||
| AK016035.1 | - | 1 | 1000 | |||