siRNA Details
| Gene Symbol: Rabl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042499.1 | NP_001035964.1 | 8 | 1494 | ||
| GenBank mRNA | AK011280.1 | - | 8 | 1436 | ||
| AK028195.1 | - | 8 | 1488 | |||
| AK028269.1 | - | 8 | 1481 | |||
| BC050194.1 | - | 8 | 1475 | 30022531 | ||

Rabl3
Genomic Map