siRNA Details
| Gene Symbol: l7Rn6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291286.1 | NP_001278215.1 | 5 | 796 | ||
| NM_001291287.1 | NP_001278216.1 | 6 | 920 | |||
| NM_001291288.1 | NP_001278217.1 | 6 | 997 | |||
| NM_001291289.1 | NP_001278218.1 | 4 | 618 | |||
| NM_026304.3 | NP_080580.1 | 5 | 856 | |||
| GenBank mRNA | AK002306.1 | - | 5 | 838 | ||
| AK003324.1 | - | 4 | 609 | |||
| AK012166.1 | - | 5 | 822 | |||
| BC083330.1 | - | 6 | 858 | |||
| DQ196315.1 | - | 5 | 834 | |||
| DQ196316.1 | - | 5 | 834 | |||

l7Rn6
Genomic Map