siRNA Details
| Gene Symbol: Pbdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026312.5 | NP_080588.1 | 6 | 482 | ||
| GenBank mRNA | AK007655.1 | - | 6 | 464 | ||
| AK007872.1 | - | 5 | 351 | |||
| AK011610.1 | - | 6 | 446 | |||
| AK030507.1 | - | 6 | 464 | |||
| AK157432.1 | - | 2 | 3147 | |||
| BC035042.1 | - | 6 | 446 | 1224959 | ||
| BC044719.1 | - | 6 | 430 | 6394829 | ||
| BC061070.1 | - | 6 | 443 | 6594922 | ||

Pbdc1
Genomic Map