siRNA Details
| Gene Symbol: Aldh3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026316.2 | NP_080592.2 | 10 | 1688 | ||
| GenBank mRNA | AF362571.1 | - | 9 | 1610 | ||
| AK004004.1 | - | 3 | 707 | |||
| AK005615.1 | - | 4 | 818 | |||
| AK154500.1 | - | 10 | 1734 | |||
| AK159447.1 | - | 10 | 1702 | |||
| AK160106.1 | - | 10 | 1703 | |||
| BC046597.1 | - | 10 | 1688 | 6312211 | ||
| BC082792.1 | - | 3 | 726 | |||

Aldh3b1
Genomic Map