siRNA Details
| Gene Symbol: Msra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253712.1 | NP_001240641.1 | 5, 6 | 725 | ||
| NM_001253714.1 | NP_001240643.1 | 4, 5 | 642 | |||
| NM_001253715.1 | NP_001240644.1 | 4, 5 | 591 | |||
| NM_001253716.1 | NP_001240645.1 | 5, 6 | 453 | |||
| NM_026322.4 | NP_080598.2 | 10, 11 | 1124 | |||
| GenBank mRNA | AK002356.1 | - | 5, 6 | 600 | ||
| AK009822.1 | - | 5, 6 | 602 | |||
| AK018338.1 | - | 5, 6 | 447 | |||
| AK049714.1 | - | 5, 6 | 726 | |||
| BC014738.1 | - | 4, 5 | 550 | 4239349 | ||
| BC089311.1 | - | 10, 11 | 1124 | 6412625 | ||
| CB953198.1 | - | 4, 5 | 513 | |||

Msra
Genomic Map