siRNA Details
Gene Symbol: Lce1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026335.2 | NP_080611.1 | 2 | 562 | ||
GenBank mRNA | AK009424.1 | - | 2 | 562 | ||
AK010064.1 | - | 2 | 563 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lce1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026335.2 | NP_080611.1 | 2 | 562 | ||
GenBank mRNA | AK009424.1 | - | 2 | 562 | ||
AK010064.1 | - | 2 | 563 |