siRNA Details
| Gene Symbol: Plgrkt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026362.2 | NP_080638.1 | 4 | 289 | ||
| GenBank mRNA | AK017171.1 | - | 4 | 313 | ||
| AK017215.1 | - | 4 | 290 | |||
| AK168454.1 | - | 4 | 265 | |||
| AK170700.1 | - | 5 | 363 | |||
| AK219906.1 | - | 3 | 227 | |||
| BC024953.1 | - | 4 | 248 | 4951101 | ||

Plgrkt
Genomic Map