siRNA Details
| Gene Symbol: Caap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026368.2 | NP_080644.2 | 1, 2 | 343 | ||
| GenBank mRNA | AK013385.1 | - | 1, 2 | 313 | ||
| AK032696.1 | - | 1, 2 | 313 | |||
| AK046647.1 | - | 1, 2 | 293 | |||
| AK076487.1 | - | 1, 2 | 298 | |||
| AK140382.1 | - | 1, 2 | 344 | |||
| AK150311.1 | - | 2, 3 | 253 | |||
| AK160297.1 | - | 1, 2 | 344 | |||
| AK160572.1 | - | 1, 2 | 344 | |||
| AK171141.1 | - | 1, 2 | 344 | |||
| BC020067.1 | - | 1, 2 | 331 | 3991510 | ||

Caap1
Genomic Map