siRNA Details
| Gene Symbol: Ahctf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026375.2 | NP_080651.2 | 2, 3 | 799 | ||
| GenBank mRNA | AB059278.1 | - | 2, 3 | 455 | ||
| BC138343.1 | - | 2, 3 | 442 | 8861363 | ||
| BC138344.1 | - | 2, 3 | 442 | 8861364 | ||

Ahctf1
Genomic Map