siRNA Details
| Gene Symbol: Snx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026386.1 | NP_080662.1 | 3, 4 | 406 | ||
| GenBank mRNA | AK002692.1 | - | 3, 4 | 406 | ||
| AK005470.1 | - | 3, 4 | 418 | |||
| AK010572.1 | - | 3, 4 | 403 | |||
| AK075929.1 | - | 3, 4 | 403 | |||
| AK145840.1 | - | 3, 4 | 457 | |||
| AK153525.1 | - | 3, 4 | 403 | |||
| BC006960.1 | - | 3, 4 | 406 | 2812557 | ||
| CT010229.1 | - | 3, 4 | 391 | |||

Snx2
Genomic Map