siRNA Details
| Gene Symbol: Wdr83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026399.2 | NP_080675.2 | 3, 4 | 472 | ||
| GenBank mRNA | AK005790.1 | - | 3, 4 | 412 | ||
| AK020654.1 | - | 3, 4 | 433 | |||
| AY365434.1 | - | 3, 4 | 472 | |||
| BC005587.1 | - | 3 | 711 | |||
| BC019369.1 | - | 3, 4 | 395 | 4014928 | ||
| BC020047.1 | - | 1314 | ||||
| BC034842.1 | - | 3 | 640 | |||

Wdr83
Genomic Map