siRNA Details
| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026417.4 | NP_080693.2 | 6 | 2079 | ||
| GenBank mRNA | AK083301.1 | - | 6 | 2059 | ||
| AK086213.1 | - | 785 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yipf4| Gene Symbol: Yipf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026417.4 | NP_080693.2 | 6 | 2079 | ||
| GenBank mRNA | AK083301.1 | - | 6 | 2059 | ||
| AK086213.1 | - | 785 | ||||