siRNA Details
| Gene Symbol: Coq10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026424.3 | NP_080700.3 | 1, 2 | 229 | ||
| GenBank mRNA | AK006551.1 | - | 1, 2 | 230 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coq10b| Gene Symbol: Coq10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026424.3 | NP_080700.3 | 1, 2 | 229 | ||
| GenBank mRNA | AK006551.1 | - | 1, 2 | 230 | ||