siRNA Details
| Gene Symbol: Cmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026442.3 | NP_080718.1 | 2 | 258 | ||
| GenBank mRNA | AK002667.1 | - | 2 | 155 | ||
| AK008567.1 | - | 2 | 177 | |||
| AK011760.1 | - | 2 | 159 | |||
| AK020626.1 | - | 2 | 142 | |||
| AK168728.1 | - | 2 | 205 | |||
| BC048547.1 | - | 2 | 239 | |||
| BC056653.1 | - | 2 | 132 | 5068125 | ||
| CJ170455.1 | - | 2 | 259 | |||

Cmc1
Genomic Map