siRNA Details
| Gene Symbol: Galntl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026449.3 | NP_080725.2 | 2 | 321 | ||
| GenBank mRNA | AK005605.1 | - | 2 | 342 | ||
| AK006193.1 | - | 2 | 300 | |||
| AK016415.1 | - | 2 | 322 | |||
| BC049554.1 | - | 2 | 303 | |||
| BC100447.1 | - | 2 | 174 | |||
| BC145757.1 | - | 2 | 181 | 40131060 | ||

Galntl5
Genomic Map