siRNA Details
| Gene Symbol: Slc48a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026353.4 | NP_080629.1 | 3 | 837 | ||
| GenBank mRNA | AB041659.1 | - | 3 | 2035 | ||
| AK002729.1 | - | 3 | 780 | |||
| AK007888.1 | - | 3 | 786 | |||
| AK088485.1 | - | 3 | 760 | |||
| AK089421.1 | - | 3 | 838 | |||
| AK138729.1 | - | 3 | 773 | |||
| AK150552.1 | - | 3 | 786 | |||
| AK155824.1 | - | 3 | 824 | |||
| AK159879.1 | - | 3 | 803 | |||
| AK159892.1 | - | 3 | 803 | |||
| AK159898.1 | - | 3 | 756 | |||
| AK168636.1 | - | 3 | 769 | |||
| AK170393.1 | - | 3 | 814 | |||
| AK181240.1 | - | 1 | 310 | |||
| AK184893.1 | - | 1 | 309 | |||
| AK189427.1 | - | 1 | 245 | |||
| AK189571.1 | - | 1 | 247 | |||
| AK193857.1 | - | 1 | 308 | |||
| AK204097.1 | - | 1 | 185 | |||
| AK213892.1 | - | 1 | 309 | |||
| AK219716.1 | - | 1 | 180 | |||
| BC022913.1 | - | 3 | 783 | 4237364 | ||

Slc48a1
Genomic Map